MAN412119132 Manchester Organics

Di-tert-butyl chloromethyl phosphate

  • MDL number MFCD12165895
  • Molecular Weight : 258.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A41217
Availability details for
Quantity
  • Availability
  • Add to Cart