MAN412119170 Manchester Organics

2,2-Difluoro-1,3-dimethyl-imidazolidine

  • MDL number MFCD03788478
  • Molecular Weight : 136.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N41252
Availability details for
Quantity
  • Availability
  • Add to Cart