MAN412119180 Manchester Organics

4-Methylsulfonylpiperidine

  • MDL number MFCD11043260
  • Cas Number : 290328-55-1
  • Molecular Weight : 163.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y41261
Availability details for
Quantity
  • Availability
  • Add to Cart