MAN412119238 Manchester Organics

4-(1-Fluoro-1-methyl-ethyl)-1H-pyrimidin-6-one

  • MDL number MFCD24604646
  • Cas Number : 240414-09-9
  • Molecular Weight : 156.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/A12271
Availability details for
Quantity
  • Availability
  • Add to Cart