MAN412119252 Manchester Organics

4,4,4-Trifluorobutan-2-one

  • MDL number MFCD00077601
  • Cas Number : 2366-70-3
  • Molecular Weight : 126.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/R29114
Availability details for
Quantity
  • Availability
  • Add to Cart