MAN412119314 Manchester Organics

Disodium 9,10-dioxoanthracene-2,7-disulfonate

  • MDL number MFCD00053255
  • Molecular Weight : 412.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 80
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N41390
Availability details for
Quantity
  • Availability
  • Add to Cart