MAN412119603 Manchester Organics

3-Acetyl-3-fluoro-tetrahydrofuran-2-one

  • MDL number MFCD00215829
  • Cas Number : 63141-03-7
  • Molecular Weight : 146.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-acetyl-3-fluorodihydro-2(3H)-furanone
Inchi Code 1S/C6H7FO3/c1-4(8)6(7)2-3-10-5(6)9/h2-3H2,1H3
Inchi Key YJBYWKDEVMXEGG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/C14238
Availability details for
Quantity
  • Availability
  • Add to Cart