MAN412119667 Manchester Organics

2-Bromo-1-(o-tolyl)ethanone

  • MDL number MFCD04038955
  • Cas Number : 51012-65-8
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A41861
Availability details for
Quantity
  • Availability
  • Add to Cart