MAN412119691 Manchester Organics

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridin-1-yl]ethanone

  • MDL number MFCD18427628
  • Cas Number : 1227068-67-8
  • Molecular Weight : 251.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-acetyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
Inchi Code 1S/C13H22BNO3/c1-10(16)15-8-6-11(7-9-15)14-17-12(2,3)13(4,5)18-14/h6H,7-9H2,1-5H3
Inchi Key RENBVEOCTVQABH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z41883
Availability details for
Quantity
  • Availability
  • Add to Cart