MAN412119692 Manchester Organics

2-(2-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD05663874
  • Cas Number : 876062-39-4
  • Molecular Weight : 222.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/A41884
Availability details for
Quantity
  • Availability
  • Add to Cart