MAN412119706 Manchester Organics

[4-(1-Hydroxy-1-methyl-ethyl)phenyl]boronic acid

  • MDL number MFCD09992905
  • Cas Number : 886593-45-9
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1-hydroxy-1-methylethyl)phenylboronic acid
Inchi Code 1S/C9H13BO3/c1-9(2,11)7-3-5-8(6-4-7)10(12)13/h3-6,11-13H,1-2H3
Inchi Key DPIDNFWCDMXMDY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/R41898
Availability details for
Quantity
  • Availability
  • Add to Cart