MAN412119708 Manchester Organics

(1-tert-Butoxycarbonylindol-3-yl)boronic acid

  • MDL number MFCD06656257
  • Molecular Weight : 261.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T41900
Availability details for
Quantity
  • Availability
  • Add to Cart