MAN412119794 Manchester Organics

5-Formylfuran-2-carboxylic acid

  • MDL number MFCD00020924
  • Cas Number : 13529-17-4
  • Molecular Weight : 140.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-formyl-2-furoic acid
Inchi Code 1S/C6H4O4/c7-3-4-1-2-5(10-4)6(8)9/h1-3H,(H,8,9)
Inchi Key SHNRXUWGUKDPMA-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z41975
Availability details for
Quantity
  • Availability
  • Add to Cart