MAN412119800 Manchester Organics

1H-1,3,5-Triazine-2,4-dione

  • MDL number MFCD07367658
  • Cas Number : 71-33-0
  • Molecular Weight : 113.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/F41981
Availability details for
Quantity
  • Availability
  • Add to Cart