MAN412119806 Manchester Organics

Methyl 5-formylpyrazine-2-carboxylate

  • MDL number MFCD12755910
  • Molecular Weight : 166.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R41990
Availability details for
Quantity
  • Availability
  • Add to Cart