MAN412119916 Manchester Organics

2-Methylpyridine-4-carbonitrile

  • MDL number MFCD06200207
  • Cas Number : 2214-53-1
  • Molecular Weight : 118.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/B42092
Availability details for
Quantity
  • Availability
  • Add to Cart