MAN424065775 Manchester Organics

5-(bromomethyl)-1-methyl-benzotriazole

  • MDL number MFCD03407338
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W47883

Safety Information

MSDS https://www.manchesterorganics.com/W47883
Availability details for
Quantity
  • Availability
  • Add to Cart