MAN424065778 Manchester Organics

4-Amino-2,5-dichloro-phenol

  • MDL number MFCD09743861
  • Molecular Weight : 178.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z47955

Safety Information

MSDS https://www.manchesterorganics.com/Z47955
Availability details for
Quantity
  • Availability
  • Add to Cart