MAN424065780 Manchester Organics

Methyl 3-amino-5-chloro-2-hydroxy-benzoate

  • MDL number MFCD03768414
  • Molecular Weight : 201.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B47474

Safety Information

MSDS https://www.manchesterorganics.com/B47474
Availability details for
Quantity
  • Availability
  • Add to Cart