MAN424065798 Manchester Organics

[5-(Prop-1-yn-1-yl)pyridin-3-yl]boronic acid

  • MDL number MFCD18258851
  • Molecular Weight : 160.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T35828
Availability details for
Quantity
  • Availability
  • Add to Cart