MAN424065837 Manchester Organics

4-(Oxetan-3-yl)piperidine Hemioxalate

  • MDL number MFCD23105786
  • Molecular Weight : 372.45647

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D34389

Safety Information

MSDS https://www.manchesterorganics.com/D34389
Availability details for
Quantity
  • Availability
  • Add to Cart