MAN424065894 Manchester Organics

2,4-Dichloro-6-(trifluoromethyl)pyridine

  • MDL number MFCD10698621
  • Molecular Weight : 215.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y14731
Availability details for
Quantity
  • Availability
  • Add to Cart