MAN424065917 Manchester Organics

2-Methyl-5-(trifluoromethyl)pyrazole-3-carboxylic acid

  • MDL number MFCD02090853
  • Molecular Weight : 194.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/X42180
Availability details for
Quantity
  • Availability
  • Add to Cart