MAN424065938 Manchester Organics

4-Bromo-1,5-dimethyl-pyrazole-3-carboxylic acid

  • MDL number MFCD02090877
  • Cas Number : 5775-91-7
  • Molecular Weight : 219.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/V42201
Availability details for
Quantity
  • Availability
  • Add to Cart