MAN424066061 Manchester Organics

3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

  • MDL number MFCD12923417
  • Cas Number : 944401-75-6
  • Molecular Weight : 238.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N42333
Availability details for
Quantity
  • Availability
  • Add to Cart