MAN424066161 Manchester Organics

1H-Imidazole-4-carbonitrile

  • MDL number MFCD00233358
  • Molecular Weight : 93.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U42430
Availability details for
Quantity
  • Availability
  • Add to Cart