MAN424066339 Manchester Organics

5-(2,4-Dichlorophenyl)isoxazole-3-carboxylic acid

  • MDL number MFCD04969804
  • Molecular Weight : 258.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L42699

Safety Information

MSDS https://www.manchesterorganics.com/L42699
Availability details for
Quantity
  • Availability
  • Add to Cart