MAN424066350 Manchester Organics

3-(1-Methylpyrazol-4-yl)propanoic acid

  • MDL number MFCD04969921
  • Molecular Weight : 154.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y43055

Safety Information

MSDS https://www.manchesterorganics.com/Y43055
Availability details for
Quantity
  • Availability
  • Add to Cart