MAN424066353 Manchester Organics

2-(3-Nitro-1,2,4-triazol-1-yl)acetic acid

  • MDL number MFCD00297307
  • Molecular Weight : 172.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B42644

Safety Information

MSDS https://www.manchesterorganics.com/B42644
Availability details for
Quantity
  • Availability
  • Add to Cart