MAN424066362 Manchester Organics

5,7-Dimethylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD01051014
  • Molecular Weight : 191.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L42722

Safety Information

MSDS https://www.manchesterorganics.com/L42722
Availability details for
Quantity
  • Availability
  • Add to Cart