MAN424066365 Manchester Organics

2-(4-Ethoxyphenoxy)propanoic acid

  • MDL number MFCD03419317
  • Molecular Weight : 210.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P42771

Safety Information

MSDS https://www.manchesterorganics.com/P42771
Availability details for
Quantity
  • Availability
  • Add to Cart