MAN424066368 Manchester Organics

5-(1-Piperidylmethyl)furan-2-carboxylic acid

  • MDL number MFCD00455269
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T42866

Safety Information

MSDS https://www.manchesterorganics.com/T42866
Availability details for
Quantity
  • Availability
  • Add to Cart