MAN424066371 Manchester Organics

5-(1,3-Dimethylpyrazol-4-yl)isoxazole-3-carboxylic acid

  • MDL number MFCD04970278
  • Molecular Weight : 207.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W43007

Safety Information

MSDS https://www.manchesterorganics.com/W43007
Availability details for
Quantity
  • Availability
  • Add to Cart