MAN424066373 Manchester Organics

1-(Difluoromethyl)-5-methyl-pyrazole-3-carboxylic acid

  • MDL number MFCD04970128
  • Molecular Weight : 176.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y43078

Safety Information

MSDS https://www.manchesterorganics.com/Y43078
Availability details for
Quantity
  • Availability
  • Add to Cart