MAN424066380 Manchester Organics

5-(4-Hydroxyphenyl)isoxazole-3-carboxylic acid

  • MDL number MFCD05237205
  • Molecular Weight : 205.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F42717

Safety Information

MSDS https://www.manchesterorganics.com/F42717
Availability details for
Quantity
  • Availability
  • Add to Cart