MAN424066394 Manchester Organics

Ethyl 6-methoxy-1H-indole-2-carboxylate

  • MDL number MFCD01544121
  • Molecular Weight : 219.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W43030

Safety Information

MSDS https://www.manchesterorganics.com/W43030
Availability details for
Quantity
  • Availability
  • Add to Cart