MAN424066396 Manchester Organics

3-Bromoquinolin-6-amine

  • MDL number MFCD07757603
  • Molecular Weight : 223.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y43101

Safety Information

MSDS https://www.manchesterorganics.com/Y43101
Availability details for
Quantity
  • Availability
  • Add to Cart