MAN424066406 Manchester Organics

3-(2-Methoxyphenyl)-4,5-dihydroisoxazole-5-carboxylic acid

  • MDL number MFCD06011105
  • Molecular Weight : 221.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J42743

Safety Information

MSDS https://www.manchesterorganics.com/J42743
Availability details for
Quantity
  • Availability
  • Add to Cart