MAN424066414 Manchester Organics

1,3,6-Trimethylpyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD06739303
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T42912

Safety Information

MSDS https://www.manchesterorganics.com/T42912
Availability details for
Quantity
  • Availability
  • Add to Cart