MAN424066421 Manchester Organics

2-(3,4-Dichloroanilino)acetic acid

  • MDL number MFCD02255628
  • Molecular Weight : 220.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A42712

Safety Information

MSDS https://www.manchesterorganics.com/A42712
Availability details for
Quantity
  • Availability
  • Add to Cart