MAN424066430 Manchester Organics

2-(4-Hydroxyphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD00687527
  • Molecular Weight : 265.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K42767

Safety Information

MSDS https://www.manchesterorganics.com/K42767
Availability details for
Quantity
  • Availability
  • Add to Cart