MAN424066436 Manchester Organics

2-(2-Pyridylsulfanyl)acetic acid

  • MDL number MFCD00276114
  • Molecular Weight : 169.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S42888

Safety Information

MSDS https://www.manchesterorganics.com/S42888
Availability details for
Quantity
  • Availability
  • Add to Cart