MAN424066449 Manchester Organics

2-[3-(Tetrazol-1-yl)phenoxy]acetic acid

  • MDL number MFCD02598026
  • Molecular Weight : 220.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F42786

Safety Information

MSDS https://www.manchesterorganics.com/F42786
Availability details for
Quantity
  • Availability
  • Add to Cart