MAN424066451 Manchester Organics

2-(4-Nitrophenoxy)propanoic acid

  • MDL number MFCD03422224
  • Molecular Weight : 211.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H42788

Safety Information

MSDS https://www.manchesterorganics.com/H42788
Availability details for
Quantity
  • Availability
  • Add to Cart