MAN424066464 Manchester Organics

2-[3-(Trifluoromethyl)-4,5,6,7-tetrahydroindazol-1-yl]acetic acid

  • MDL number MFCD02732814
  • Molecular Weight : 248.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X43146

Safety Information

MSDS https://www.manchesterorganics.com/X43146
Availability details for
Quantity
  • Availability
  • Add to Cart