MAN424066475 Manchester Organics

4-Oxo-3H-quinazoline-7-carboxylic acid

  • MDL number MFCD03942331
  • Cas Number : 202197-73-7
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/J42743
Availability details for
Quantity
  • Availability
  • Add to Cart