MAN424066495 Manchester Organics

4-Ethyl-5-methyl-thiophene-3-carboxylic acid

  • MDL number MFCD03419845
  • Molecular Weight : 170.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F42832

Safety Information

MSDS https://www.manchesterorganics.com/F42832
Availability details for
Quantity
  • Availability
  • Add to Cart