MAN424066498 Manchester Organics

4-(2,2,2-Trifluoroethoxy)benzoic acid

  • MDL number MFCD05669500
  • Molecular Weight : 220.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J42835

Safety Information

MSDS https://www.manchesterorganics.com/J42835
Availability details for
Quantity
  • Availability
  • Add to Cart