MAN424066513 Manchester Organics

3-[(5-Methyl-3-nitro-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD00849832
  • Molecular Weight : 261.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A42804

Safety Information

MSDS https://www.manchesterorganics.com/A42804
Availability details for
Quantity
  • Availability
  • Add to Cart