MAN424066519 Manchester Organics

2-(2,4-Difluorophenoxy)acetic acid

  • MDL number MFCD03422203
  • Molecular Weight : 188.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G42856

Safety Information

MSDS https://www.manchesterorganics.com/G42856
Availability details for
Quantity
  • Availability
  • Add to Cart